Tuberculin syringes, with slip tip, BD™
Catalog # BDAM309659
Supplier: BD Medical
Specifications
- Capacity:1 ml
- Description:Disposable tuberculin syringe with slip tip
- Seq:01
- Pk:200
Specifications
About this item
BD™ Tuberculin syringe features bold, clear scale markings to assist in making easy and accurate dosage measurements. The thinner and smaller syringe size is easier to control and is less threatening to the patient.