Order Entry
China
Orders LinkContactUsLinkComponent
VWR® Talon® , Lab frames, small

VWR® Talon® , Lab frames, small

Lab frame with aluminum rods, 610 mm
Catalog # VWRU60075-008
VWR® Talon® , Lab frames, small
Catalog # VWRU60075-008
VWR® Talon® , Lab frames, small
VWR® Talon® , Lab frames, small

Lab frame with aluminum rods, 610 mm

Price
¥9,280.00
Log in for availability
1 * 1 件
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

These vertical mounting lab frames are ideal for glassware set up in small laboratories or where space is limited. Frames measure 610×610 mm (24×24") with a 457 mm (18") wide base for stability. Base may be permanently mounted to benchtop with screws (included).

Base dimension: 457 mm

Frame dimensions: 610×610 mm

 : Each lab frame includes eight 51 mm rods, two 457 mm rods, eight 610 mm rods, four rod end connectors, eight S connectors and four lab frame feet.

Specifications
  • Description:
    Lab frame with aluminum rods
  • Length:
    610 mm

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.