VWR®, Ring Weights for Flasks, C-Shaped
Supplier: VWR BRAND PRIVATE LABEL USA
Certificates
About this item
Lead core, with Vikem® vinyl coating. These lead rings fit neatly around the neck of filtering flasks with side-arm tubing connections without disconnecting the tubing.
- Stabilise vessels in water baths or on the benchtop
- Open ring is flexible enough to be pulled apart to accommodate a wider vessel