Order Entry
China
Orders LinkContactUsLinkComponent
VWR®, Ring Weights for Flasks
VWR®, Ring Weights for Flasks
VWR®, Ring Weights for Flasks
VWR®, Ring Weights for Flasks
Vikem® vinyl coated lead rings prevent flasks floating during water bath incubations and help prevent spillages or breakages.
  • Bright red vinyl covering protects flask from scratches
  • Stabilise flasks in water baths or on the benchtop
Product Family Options
  • For
    Ø ext.
    Ø int.
    Weight
  • Flasks 125 - 500 ml
    76 mm
    43 mm
    200 g
    1 * 1 件
    1 * 1 件 - ¥790.00
    ¥790.00
    1 * 1 件
       ¥790.00
  • Flasks 250 - 1000 ml
    89 mm
    51 mm
    500 g
    1 * 1 件
    1 * 1 件 - ¥848.00
    ¥848.00
    1 * 1 件
       ¥848.00
  • Flasks 500 - 2000 ml
    102 mm
    56 mm
    700 g
    1 * 1 件
    1 * 1 件 - ¥1,004.00
    ¥1,004.00
    1 * 1 件
       ¥1,004.00
  • Flasks 1000 ml or larger
    114 mm
    71 mm
    900 g
    1 * 1 件
    1 * 1 件 - ¥1,078.00
    ¥1,078.00
    1 * 1 件
       ¥1,078.00