VWR®TraceClean®, Water sampling bottles, hdpe, wide neck
Catalog # VWRU89094-094
Supplier: VWR BRAND PRIVATE LABEL USA
Specifications
- Capacity:250 ml
- Description:Not Processed (NPC) HDPE bottle with poly-foam lined cap
- Seq:01
- Environmentally Preferable:
- Pk:48
Specifications
About this item
HDPE, translucent.
- Available with poly-foam lined caps
- For use in collecting clays, slurries, and sediment matrices for use in geotechnical, environmental, toxicity characteristic leaching procedure (TCLP) metals, inorganic compounds, and benthic organism studies
Processed (PC) and Quality-Assured® (QA) bottles follow EPA cleaning procedures listed in the OSWER Directive 9240.0-05A 'Specifications and Guidance for Contaminant-Free Sample Containers.' QA packers include a certificate of analysis in every case. QA packers also include bar codes with lot number and individual container number for complete traceability. Not Processed (NPC) packers do not receive the specialty cleaning process. All cases are custody-sealed to ensure content integrity.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904