VWR®, Bulbs for Pipettes
Supplier: VWR BRAND PRIVATE LABEL USA
Certificates
About this item
Pipette bulbs made of natural rubber.
- Durable material
- Bulbs are suitable for medicine droppers and small dropping pipettes
- Neck is grooved to take flat flange of pipettes
- Heavy-walled, cylindrical in shape
- Bulbs fit pipettes 6 to 8 mm Ext.Ø