VWR® Multistage gas regulators feature safe, accurate and sensitive delivery of reduced pressure. Assembly features include: high-pressure valve, low-pressure valve, intermediate pressure relief valve between the two stages, back cap vent holes and bonnet vent holes. High and low pressure PTFE encapsulated seat. Regulator damage from contaminants is minimal.
- Solderless construction for easy maintenance
- Solid forged brass body contains two regulators
- First stage reduces full cylinder pressure to 90%
- Sintered inlet filter removes particulates, tank scale and rust from gas stream
- Stainless steel diaphragms
Note: For non corrosive gases only.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Catalog No:
- VWRU55850-412
- VWRU55850-422
- VWRU55850-414
- Type:
- Multi stage
- Multi stage
- Multi stage
- Max. inlet pressure:
- 275,80 bar
- 275,80 bar
- 275,80 bar
- Outlet pressure:
- 0 - 6,44 bar
- 0 - 6,44 bar
- 0 - 6,44 bar
- Flow:
- 7,22/8,64/68,81 m³/h
- 7,22/8,64/68,81 m³/h
- 7,22/8,64/68,81 m³/h