Order Entry
China
ContactUsLinkComponent
Tetramethylammonium hydroxide 10% in aqueous solution, BAKER ANALYZED®, J.T.Baker®
Toxic
Corrosive
Tetramethylammonium hydroxide 10% in aqueous solution, BAKER ANALYZED®, J.T.Baker®
Catalog # BAKRV643-07
Supplier:  Avantor
CAS Number:  
undefined
Toxic
Corrosive
Tetramethylammonium hydroxide 10% in aqueous solution, BAKER ANALYZED®, J.T.Baker®
Catalog # BAKRV643-07
Supplier:  Avantor
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle, clear in a single shipper
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

Specification Test Results

  • Assay ((CH₃)₄NOH):
    ≥9.8 %
  • Residue after Ignition:
    ≤0.020 %
  • Ammonia and Other Amines:
    Passes Test

Specification Test Results

About this item

 :
 :91.15 g/mol
Boiling Pt:110 °C (1013 hPa)
Density:1.02 g/cm³ (20 °C)
ADR:8,II
 :Management of Change (MOC) category = N
EINECS:200-882-9
UN:1835
 :GHS06
 :GHS05
Storage Temperature:Ambient
CAS Number:75-59-2
Formula:C₄H₁₃NO
MDL Number:MFCD00008280