Water with 0,1% (v/v) formic acid, BAKER ANALYZED® for LC-MS, J.T.Baker®
Supplier: Avantor
Certificates
About this item
:7732-18-5
Storage Temperature:Ambient
EINECS:200-579-1
REACH:01-2119491174-37-XXXX
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:Passes Test
- Appearance:Passes Test
- Formic Acid:0.095 - 0.105 %
- Gradient Elution Test (a.u.) - 254 nm:≤ 0.01
- Residue after Evaporation:≤ 1.0 ppm
- LC/MS Suitability - Largest Response on ESI- Positive Mode (as Reserpine):≤ 50 ppb
- Trace Impurities - Aluminum (Al):≤ 50.0 ppb
- Trace Impurities - * Calcium (Ca):≤ 50.0 ppb
- Trace Impurities - Iron (Fe):≤ 50.0 ppb
- Trace Impurities - Magnesium (Mg):≤ 50.0 ppb
- Trace Impurities - Potassium (K):≤ 50 ppb
- Trace Impurities - * Sodium (Na):≤ 50.0 ppb