VWR®, Measuring Cylinders, Low Form, Class B, Borosilicate Glass
Borosilicate glass, low form, class B.
- Calibrated to deliver (TD)
- Two pour outs are provided
- Graduations printed in durable white ceramic enamel
: Glass conforms to USP Type I and ASTM E438, Type I, class A requirements.
: Supplied with an LDPE SAFE-GARD® bumper.