Order Entry
China
ContactUsLinkComponent
VWR®, Pipette Filler
VWR®, Pipette Filler
Catalog # VWRU53502-205
undefined
VWR®, Pipette Filler
Catalog # VWRU53502-205

Specifications

  • Colour:
    Red
  • Description:
    Pipette filler
  • Pk:
    1

Specifications

About this item

Pipette bulb made of synthetic rubber.

  • For use with pipettes from 2 to 50 ml
  • Made of synthetic rubber with glass ball valves
  • No metal parts to corrode
  • Supplied with an adapter for capacities greater than 20 ml