Order Entry
China
ContactUsLinkComponent
VWR® Traceable® Humidity Sponge™, Dessicants
  VWRU61161-321
undefined
VWR® Traceable® Humidity Sponge™, Dessicants
  VWRU61161-321
  • Type:
    Non-toxic desiccant
  • Description:
    Humidity Sponge™ Non-indicating, can be regenerated for reuse
  • Seq:
    01
  • W×D×H:
    76,2×76,2×6,4 mm
  • Pk:
    40
Humidity Sponges™ meet FDA (Food and Drug Administration) requirements for use in direct contact with food and drugs. Desiccant (sodium calcium aluminosilicate hydrate) is non toxic and is a GRAS (Generally Recognised as Safe) material. It is ideal for storing food/pharmaceutical samples, electronics, PC boards, optics, chemicals and equipment. Outer material is made from DuPont’s Tyvek® breathable, high-density polyethylene.
  • Drying capability depends on volume, temperature, and moisture
  • Bags are guaranteed to retain 16% moisture by weight, typical results are 37% moisture absorption
  • Indicator sponge shows when moisture capacity has been reached
 : Meets FDA requirements—certified ISO Class 5 (Class M3.5) cleanroom-compatible.