Stoppers, Standard Taper, HDPE, PYREX®
Catalog # CORN7624-32
Supplier: CORNING SCIENCE PRODUCTS
Specifications
- Ø bottom:32,0 mm
- Seq:10
- Pk:6
Specifications
About this item
These PYREX® No.13 stoppers are made from linear HDPE and have closed bottoms to conform to Standard Taper stopper dimensions.
- Blue and white with a closed bottom
- Three rings on the plug provide an efficient seal
- Improved design fully protects the neck from accidental tipping