Avantor® ACE®, Guard Cartridge Holders
: Avantor
Stainless steel, Avantor® ACE® guard cartridge holders are used for connecting guard cartridges to the LC column which then provides column protection and can extend the lifetime of your column. Holders are required for all Avantor® ACE® guard cartridges. Stand-alone holders require the use of a suitable connector often known as a column coupler.
- Catalog No:
- ACETH0001
- ACEUBIO-GRD-0001
- ACETH0002
- ACETH0011
- ACETH0009
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)