Order Entry
China
ContactUsLinkComponent
Avantor® ACE®, HPLC ¹/₃₂" Column Connectors, Finger-Tight
Avantor® ACE®, HPLC ¹/₃₂" Column Connectors, Finger-Tight
Catalog # ACETACE-MC3210
Supplier:  Avantor
   Low Manufacturing Impact
undefined
Avantor® ACE®, HPLC ¹/₃₂" Column Connectors, Finger-Tight
Catalog # ACETACE-MC3210
Supplier:  Avantor
   Low Manufacturing Impact
Specifications
  • Description:
    Microbore finger-tight HPLC column connector, 1/32", 6-40 thread
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    ISO 14001:2015 (530840 UM15)
  • Pk:
    10
Specifications
About this item
PEEK ¹/₃₂" microbore HPLC column inlet / outlet connectors. These column connectors are recommended for ¹/₃₂" microbore HPLC column connections. They are compatible with both capillary (360 µm OD) and ¹/₃₂" connection tubing.
  • Pressure rated to 400 bar (6000 psi)
  • Reusable
  • Compatible with all microbore columns with ¹/₃₂" and 6-40 threads