Avantor® ACE® C18-300, HPLC Columns, 5 µm
Supplier: Avantor
Certificates
About this item
Avantor® ACE® C18-300 columns provide great reproducibility and column lifetime. These stainless steel columns are available in a wide range of particle sizes and dimensions from capillary to preparative. These columns are designed for a wide range of chromatographic applications, to provide great performance with acidic, basic and neutral molecules.
- Designed for peptide and protein separations
- Ultra high purity, base deactivated silica with exceptional reproducibility
- Ideal for a wide range of analytes differing in hydrophobicity
- Wide range of particle sizes (3, 5 and 10 µm) and complementary phases
- Method validation kits contain three different batches of the same phase - available on request
These columns provide excellent peak shape even at extremely low levels of additives such as TFA, making them ideal for LC/MS applications.
Specifications
- Catalog No:
- ACETACE-221-2546
- ACETACE-221-1202
- ACETACE-221-2510
- ACETACE-221-1046
- ACETACE-221-2520
- ACETACE-221-2504
- ACETACE-221-1010
- ACETACE-221-1020
- ACETACE-221-0502
- ACETACE-221-0503
- ACETACE-221-1220
- ACETACE-221-2502
- ACETACE-221-2503
- ACETACE-221-7546
- ACETACE-221-1520
- ACETACE-221-1546
- ACETACE-221-1002
- ACETACE-221-0520
- ACETACE-221-1510
- ACETACE-221-0546
- ACETACE-221-7520
- ACETACE-221-0103GD
- ACETACE-221-1503
- ACETACE-221-0102GD
- ACETACE-221-1504
- ACETACE-221-0510
- ACETACE-221-7502
- ACETACE-221-1502
- ACETACE-221-0110GD
- ACETACE-221-1008
- ACETACE-221-3046
- ACETACE-221-1003
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Carbon load:
- 9,0%
- 9,0%
- 9,0%
- 9,0%
- 9,0%
- 9,0%
- 9,0%
- 9,0%
- 9,0%
- 9,0%
- 9,0%
- 9,0%
- 9,0%
- 9,0%
- 9,0%
- 9,0%
- 9,0%
- 9,0%
- 9,0%
- 9,0%
- 9,0%
- 9,0%
- 9,0%
- 9,0%
- 9,0%
- 9,0%
- 9,0%
- 9,0%
- 9,0%
- Endcapped:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Max. pressure:
- 275 bar
- 275 bar
- 275 bar
- 275 bar
- 275 bar
- 275 bar
- 275 bar
- 275 bar
- 275 bar
- 275 bar
- 275 bar
- 275 bar
- 275 bar
- 275 bar
- 275 bar
- 275 bar
- 275 bar
- 275 bar
- 275 bar
- 275 bar
- 275 bar
- pH range:
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- Surface area:
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- 100 m²/g
- USP code:
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- Environmental certification:
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
Specifications
Related Information
Avantor® ACE® HPLC and UHPLC Columns
Product Literature
TDS Avantor ACE® HPLC and UHPLC Columns