Order Entry
China
ContactUsLinkComponent
Library preparation kits, KAPA™ Hyper Prep Illumina
Library preparation kits, KAPA™ Hyper Prep Illumina
  KAPBKK8502
 :  
Library preparation kits, KAPA™ Hyper Prep Illumina
  KAPBKK8502
 :  
  • Pk:
    24 Tests
  • Description:
    KAPA™ Hyper Prep Illumina, 24 reactions
The KAPA™ Hyper Prep Kit is a versatile, streamlined solution for DNA library preparation for Illumina sequencing. The one-tube chemistry, optimally formulated, and evolved enzymes enable higher yields of adapter-ligated library and lower amplification bias. This translates to higher library diversity, lower duplication rates and more uniform coverage, particularly for FFPE and low-input samples.
  • Robust performance with inputs from 1 ng to 1 µg
  • High-quality libraries from 250 ng FFPE DNA or less
  • Complete library construction in less than 3 hours
  • KAPAHiFi™ DNA Polymerase for lower amplification bias
  • One-tube workflow with minimal cleanup steps
 : Kit contains the following modules: end repair, A-tailing, adaptor ligation, and library amplification. Kits without amplification reagents are also available. Adapters and SPRI cleanup beads are not supplied with this kit.