Labels, removable, ITW Alma
Catalog # TEXWALB009AAAGL
Supplier: ITW-CONTAMINATION TEXWIPE
Specifications
- Colour:White
- Description:Removable labels
- Dimensions:25×76 mm
- Material:Polyolefin
- Packed:On roll
- Pk:1.000
Specifications
About this item
Nonvinyl polyolefin labels is designed for cleanroom pens, thermal transfer and direct thermal printers.
- Peel off easily without leaving adhesive residue
- Smooth edges for low particle and contamination levels
- Adhere to the same surfaces as conventional labels
Packaging: Packaged 1000 labels per roll and supplied clean with Alma's proprietary process. Labels are wound on a PE core, double-bagged and heat-sealed.