Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
¥0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
China
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Environmental Test Kits
Accessories interference removal for Hach photometric analysis kits
Accessories interference removal for Hach photometric analysis kits
Accessories interference removal for Hach photometric analysis kits
Catalog # HACH211220
Supplier:
HACH
CAS Number:
Expand image
Accessories interference removal for Hach photometric analysis kits
Catalog # HACH211220
Supplier:
HACH
CAS Number:
Specifications
Pk:
29 mL
Description:
Phenol solution for interference removal, 30 g/litre
Specifications
About this item
Product Details
Product Details