Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody type:Primary
- Antigen symbol:H-2Kb/H-2Db
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Antigen synonyms:H-2Db
- Size:500 µg
- Pk:500 µG
Specifications
About this item
Type: Primary
Antigen: H-2Kb/H-2Db
Clonality: Monoclonal
Clone:
Conjugation: PURE
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Mouse