About this item
This portable meter provides simple and fast measurements of pH, mV and temperature values.
- SensION+ carrying cases work as complete, portable labs
- Screw-on calibration tubes and probe storage tubes simplify use in field
- Instruments are ergonomically shaped and can easily be operated single-handed
- IP 67-protected
The all-in-one kits for pH and ORP measurements make general electrochemistry testing fast and simple. Each system comes complete with everything you need to start testing.
pH kit for general use: Ideal for general aqueous applications. Supplied with gel-filled pH electrode 5050T with ceramic diaphragm and integrated temp. sensor.
pH kit for dirty samples: Ideal for high solid content and/or viscous media samples. Supplied with pH combination electrode 5051T with open circular diaphragm, solid electrolyte and integrated temp. sensor.
pH kit for difficult samples: Ideal for low ionic strength, colloidal, emulsions and cream samples. Supplied with gel-filled pH combination electrode 5052T with annular porous PTFE diaphragm and integrated temp. sensor.
pH kit for food applications: Ideal for food samples, e.g cheese, meat, fish, fruits, bread doughs. Supplied with pH combination puncture electrode 5053T with stainless steel body and integrated temp. sensor.
ORP kits: Ideal for chemical wastewater treatment, galvanic baths and general aqueous applications. Supplied with combination platinum ORP electrode 5055 with PC body and non refillable gel-electrolyte reference.
pH kits include: sensION+ pH1 meter, application specific pH electrode, sensION+ portable field carrying case, 125 ml of pH 4, 7 and10 calibration standards, colour-coded screwable calibration tubes, sample and measurement containers, electrode storage solution and user documentation.
ORP kit includes: sensION+ pH 1 meter, sensION+ 5055 ORP electrode, sensION+ portable field carrying case, 125 ml of ORP calibration standard 220 mV, screwable calibration tube, sample and measurement containers, and user documentation.
Specifications
- Catalog No:
- HACHLPV2550T97002
- HACHLPV2551T97002
- HACHLPV2555970002
- Model:
- sensION+ pH1
- sensION+ pH1
- sensION+ pH1
- pH range:
- −2...+19,99
- −2...+19,99
- −2...+19,99
- pH resolution:
- 0,01
- 0,01
- 0,01
- pH accuracy:
- ≤ 0,02
- ≤ 0,02
- ≤ 0,02
- pH calibration:
- 1 - 3 points
- 1 - 3 points
- 1 - 3 points
- Automatic buffer recognition:
- pH 2,01, 4,01, 7,00, 9,21, 10,02
- pH 2,01, 4,01, 7,00, 9,21, 10,02
- pH 2,01, 4,01, 7,00, 9,21, 10,02
- mV range:
- ±1999
- ±1999
- ±1999
- mV resolution:
- 0,1 (0 - 200 mV), 1 (rest of range)
- 0,1 (0 - 200 mV), 1 (rest of range)
- 0,1 (0 - 200 mV), 1 (rest of range)
- mV accuracy:
- ±1
- ±1
- ±1
- Temperature range:
- −20...+150 °C
- −20...+150 °C
- −20...+150 °C
- Temperature resolution:
- 0,1 °C
- 0,1 °C
- 0,1 °C
- Temperature accuracy:
- ±0,2 °C
- ±0,2 °C
- ±0,2 °C
- Temperature compensation:
- With temperature probe Pt 1000 or by keypad
- With temperature probe Pt 1000 or by keypad
- With temperature probe Pt 1000 or by keypad
- Display:
- Backlit LCD with pictograms, 128×64 pixel
- Backlit LCD with pictograms, 128×64 pixel
- Backlit LCD with pictograms, 128×64 pixel
- Power supply:
- 3 × AA alkaline batteries or rechargeable NiMH batteries
- 3 × AA alkaline batteries or rechargeable NiMH batteries
- 3 × AA alkaline batteries or rechargeable NiMH batteries
- Compliance:
- IP 67
- IP 67
- IP 67
- Languages:
- Icon based, no language needed
- Icon based, no language needed
- Icon based, no language needed
- Connections:
- MP-5
- MP-5
- MP-5
- W×D×H:
- 73×186×38 mm
- 73×186×38 mm
- 73×186×38 mm
- Weight:
- 0,3 kg
- 0,3 kg
- 0,3 kg