Order Entry
China
ContactUsLinkComponent
Cyclohexane ≥99.7%, PESTINORM® Supra Trace for organic trace analysis
Flammable
Health
Irritant
Environment
Cyclohexane ≥99.7%, PESTINORM® Supra Trace for organic trace analysis
Catalog # VWRC85385.320
Supplier:  VWR Chemicals
CAS Number:  
undefined
Flammable
Health
Irritant
Environment
Cyclohexane ≥99.7%, PESTINORM® Supra Trace for organic trace analysis
Catalog # VWRC85385.320
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • Appearance:
    Clear colourless liquid
  • Identity (IR):
    Conforms
  • Assay (calculated on anhydrous):
    Min. 99.7 %
  • Residue after evaporation:
    Max. 0.0003 %(m)
  • Water:
    Max. 0.01 M%
  • Colour:
    Max. 10 APHA
  • GC/ECD dioxins, furans and PCB's:
    Max. 5 ng/l
  • GC/ECD any pesticide (as Lindane):
    Max. 5 ng/l
  • GC/NPD any pesticide (as Parathion):
    Max. 10 ng/l
  • GC/ECD 1,2,4-TCB to deca-PCB(as Lindane):
    Max. 5 pg/ml
  • GC/ECD DCM to 1,2,4-TCB (as TCM):
    Max. 2.0 ng/ml
  • GC/MSD C10 to C40 (as Decane; 30-600amu):
    Max. 2.0 ng/ml

Specification Test Results

About this item

 :
 :84.16 g/mol
Merck Index:13,02752
Boiling Pt:81 °C (1013 hPa)
Melting Pt:7 °C
Density:0.7785 g/cm³ (20 °C)
Flash Pt:–18 °C
Storage Temperature:Ambient
EINECS:203-806-2
UN:1145
ADR:3,II
 :Filtered through a 0.2 µm filter, packaged under inert gas.
REACH:01-2119463273-41
 :GHS02
 :GHS08
 :GHS07
 :GHS09
CAS Number:110-82-7
Formula:C₆H₁₂
MDL Number:MFCD00003814