Flasks, iodine, with PTFE stopper, KIMAX®, KIMBLE®
Catalog # KIML27200-250
Supplier: KIMBLEKONTES
Specifications
- Ext.Ø:82 mm
- Capacity:250 ml
- Seq:02
- Pk:8
Specifications
About this item
Flasks, manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements are used in the analysis of dry shellac and shellac varnishes.
- Designed from ASTM Specification E1404, Type II, Class 3 requirements
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
May also be used to determine the total iodine value of oils, fatty acids, and bodied oils with Rosenmund-Kuhnhenn reagent (ASTM D1541). The 500 mL flask is required in the testing of drying oils for use in paints and varnishes, either with Wijs solution or Hanus iodine-bromine solution (ASTM D555). With [ST] 22 PTFE stopper.