Order Entry
China
ContactUsLinkComponent
Wipes for delicate tasks, KIMTECH SCIENCE* KIMWIPES*
  KIMB34705
undefined
Wipes for delicate tasks, KIMTECH SCIENCE* KIMWIPES*
  KIMB34705
  • Packed:
    15 boxes of 119 sheets
  • Colour:
    White
  • Description:
    KIMTECH SCIENCE* delicate task wipes, 2 ply
  • L×W:
    300×300 mm
  • Seq:
    01
  • Pk:
    15
This 2-ply wipe offers a unique combination of low-lint tissue softness and multi-ply absorbency. Perfect for a multitude of delicate tasks.
  • Multi-ply absorbency wipes up liquid and dust
  • Wipes clean with extra low-lint and low extractable performance
  • One-at-a-time dispensing