Test tubes, graduated, with beaded rim, KIMBLE®
KIML46350-10
- Ext.Ø:16 mm
- Capacity:10 ml
- Length:125 mm
- Pk:1
Borosilicate glass, graduated.
- With beaded top
- Scale is permanent brown stain
: Conforms to USP Type I and ASTM E438, Type I, Class A requirements.