Capillary columns, HP-PLOT Al₂O₃ S
Supplier: AGILENT HEWLETT PACKARD
Certificates
About this item
These columns are with middle range polarity. It is ideal for hydrocarbon analysis, and for resolving acetylene from butane and propylene from isobutane.
Aluminium oxide deactivated with sodium sulfate