Order Entry
China
ContactUsLinkComponent
Wipes for critical tasks, KIMTECH PREP* KIMTEX*
Wipes for critical tasks, KIMTECH PREP* KIMTEX*
Catalog # KIMB33560
Supplier:  KIMBERLY CLARK
undefined
Wipes for critical tasks, KIMTECH PREP* KIMTEX*
Catalog # KIMB33560
Supplier:  KIMBERLY CLARK
Specifications
  • Packed:
    66 sheets/pack
  • Colour:
    Blue
  • Description:
    KIMTECH PREP* KIMTEX* wipes, quarterfold format
  • L×W:
    305×318 mm
  • Seq:
    03
  • Pk:
    66
Specifications
About this item
These PP wipes are engineered for critical applications involving solvents, inks, and paints. Typical applications include: furniture finishing, engine assembly/repair, pre-packaging wipe down, LCD panel assembly, appliance assembly, printing, and glass or window cleaning.
  • Acid, base and solvent resistant: Can be used with chemicals
  • Perfect for use with lubricants, oil and grease
  • Low linting
Colour: Blue
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904