Surgical masks, PFR95* N95
Supplier: KIMBERLY CLARK
Certificates
About this item
Particulate filter respirator and surgical masks has a flat-fold duckbill design with pouch style.
- Two polyurethane headbands secure the mask to face for tight facial seal
- Soft lining and two levels of fluid protection
- Large breathing chamber for added comfort
- Provides a minimum of 95% filtration efficiency of 0,3 μm particles
Certifications: NIOSH approved