Order Entry
China
ContactUsLinkComponent
Nitrile gloves, KLEENGUARD* G10 Grey Nitrile
undefined
Nitrile gloves, KLEENGUARD* G10 Grey Nitrile
  • Catalog No:
  • KIMB97823
  • KIMB97822
  • KIMB97821
  • Model:
  • G10
  • G10
  • G10
  • Ambidextrous / hand-specific:
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Disposable / reusable:
  • Disposable
  • Disposable
  • Disposable
  • Latex / latex-free:
  • Latex-free
  • Latex-free
  • Latex-free
  • Powdered / powder-free:
  • Powder-free
  • Powder-free
  • Powder-free
  • Finish:
  • Textured fingertips
  • Textured fingertips
  • Textured fingertips
  • Cuff style:
  • Beaded
  • Beaded
  • Beaded
  • Cuff thickness:
  • 0,05 mm
  • 0,05 mm
  • 0,05 mm
  • Finger thickness:
  • 0,07 mm
  • 0,07 mm
  • 0,07 mm
  • Palm thickness:
  • 0,06 mm
  • 0,06 mm
  • 0,06 mm
  • EN norm:
  • EN 455-2
  • EN 455-2
  • EN 455-2
  • Food contact approved:
  • +
  • +
  • +