Order Entry
China
ContactUsLinkComponent
Examination gloves, nitrile, PURPLE NITRILE*
Examination gloves, nitrile, PURPLE NITRILE*
Supplier:  KIMBERLY CLARK
   Sustainable Options Available
Certificates
undefined
Examination gloves, nitrile, PURPLE NITRILE*
Supplier:  KIMBERLY CLARK
   Sustainable Options Available
Specifications
  • Catalog No:
  • KIMB55080
  • KIMB55083
  • KIMB55081
  • KIMB55082
  • Ambidextrous / hand-specific:
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Environmentally Preferable:
  • Waste Reducing
  • Waste Reducing
  • Waste Reducing
  • Disposable / reusable:
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Latex / latex-free:
  • Latex-free
  • Latex-free
  • Latex-free
  • Latex-free
  • Powdered / powder-free:
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Finish:
  • Textured fingertips
  • Textured fingertips
  • Textured fingertips
  • Textured fingertips
  • Cuff style:
  • Beaded
  • Beaded
  • Beaded
  • Beaded
  • Cuff thickness:
  • 0,09 mm
  • 0,09 mm
  • 0,09 mm
  • 0,09 mm
  • Finger thickness:
  • 0,15 mm
  • 0,15 mm
  • 0,15 mm
  • 0,15 mm
  • Palm thickness:
  • 0,12 mm
  • 0,12 mm
  • 0,12 mm
  • 0,12 mm