Order Entry
China
ContactUsLinkComponent
Anti-ATP1B1 Mouse Monoclonal Antibody [clone: M17-P5-F11]
Anti-ATP1B1 Mouse Monoclonal Antibody [clone: M17-P5-F11]
Catalog # ABCAAB2873-100
Supplier:  ABCAM
undefined
Anti-ATP1B1 Mouse Monoclonal Antibody [clone: M17-P5-F11]
Catalog # ABCAAB2873-100
Supplier:  ABCAM

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    beta Sodium Potassium ATPase
  • Clonality:
    Monoclonal
  • Clone:
    M17-P5-F11
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2a
  • Western blot:
    Yes
  • Seq:
    1286
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pack type:
    Vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [M17-P5-F11] to ATP1B1.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: ATP1B1
Clonality: Monoclonal
Clone: M17-P5-F11
Conjugation:
Epitope: This antibody recognizes an epitope between amino acid residues 195-199 of sheep sodium/potassium ATPase beta 1.
Host: Mouse
Isotype: IgG2a
Reactivity: Human;Mouse