Carboys, space saver, with screw cap, SP Bel-Art
BELAH109390005
- Capacity:5 l
- W×D×H:241×89×375 mm
- Environmentally Preferable:
- Pk:1
PP, translucent.
- In addition to the top handle, a convenient grip hold in the front facilitates removal from a shelf
- Graduated markings on front in liters
- Good chemical and temperature resistance
Space saver book bottle stacks book-like on shelves for efficient use of space.