

Trifluoroacetic acid ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:76-05-1
:
Formula:C₂HF₃O₂
MDL Number:MFCD00004169
:114.02 g/mol
Merck Index:13,09754
Boiling Pt:71.8 °C (1013 hPa)
Melting Pt:–15 °C
Density:1.490 g/cm³ (20 °C)
UN:2699
ADR:8,I
:Packaged under nitrogen.
EINECS:200-929-3
:GHS07
:GHS05
Storage Temperature:Ambient
Specification Test Results
- Assay (on anhydrous substance):Min. 99.9 %
- Evaporation residue:Max. 0.0001 %
- Water:Max. 0.005 %
- Ca (Calcium) (0.1 %):Max. 0.2 ppm
- K (Potassium) (0.1 %):Max. 0.2 ppm
- Mg (Magnesium) (0.1 %):Max. 0.2 ppm
- Na (Sodium) (0.1 %):Max. 0.5 ppm
- Transmittance (205 nm) (0.1 %):Min. 10 %
- Transmittance (220 nm) (0.1 %):Min. 50 %
- Transmittance (230 nm) (0.1 %):Min. 80 %
- Transmittance (250 nm) (0.1 %):Min. 95 %
- Transmittance (255 nm) (0.1 %):Min. 99 %
- Suitable for LC-MS:Passes test
Specification Test Results
Related Information
Certificates
Search by Lot Numberspec_VWRC84868.180 spec_VWRC84868.180 spec_VWRC84868.180 spec_VWRC84868.260 spec_VWRC84868.260
Product specification
More
VWR® LC-MS Solvents, Additives, Mixes and Associated Products