Order Entry
China
Orders LinkContactUsLinkComponent
Insulate containers, with locking clamps
Supplier:   BOCHEM
Insulate containers, with locking clamps
Supplier:  BOCHEM
Certificates
Insulate containers, with locking clamps
3 Options Available Below
Product Details & Documents
Stainless steel 18/10.
  • With three locking clamps, lid and silicone gasket
  • Valve in the lid compensates possible over and under pressure
  • Very good chemical and temperature resistance
Product Family Options
Product Information
  • Capacity
    Height
    Int.Ø
    Availability
    Price
  • 6,5 l
    160 mm
    240 mm
    Log in for availability
    Log in for availability
    1 * 1 件
    1 * 1 件
  • 10,5 l
    250 mm
    240 mm
    Log in for availability
    Log in for availability
    1 * 1 件
    1 * 1 件
  • 15,5 l
    230 mm
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 件
    1 * 1 件