Diagnostic vials, Nalgene®
Supplier: Thermo Fisher Scientific
Certificates
About this item
These clear PETG vials with white HDPE closures are ideal for sampling, storage and shipment of reagents or buffer solutions.
- Non-pyrogenic and non-cytotoxic
- Includes lined, white HDPE closure for a leakproof seal
Certifications: Manufactured in compliance with ISO 13485:2003 quality requirements. Serum Vials meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Packaging: Packed in shrink-wrapped trays with closures attached. These vials are bulk packed, 100 vials per tray, 500 vials per case.
Specifications
- Catalog No:
- NALG342035-0005
- NALG342035-0010
- NALG342035-0020
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- NAR Certificate of Retirement, ISO 14001 (Certificate Registration No. EMS 752603)
- NAR Certificate of Retirement, ISO 14001 (Certificate Registration No. EMS 752603)