VWR®, Sterile Cleanroom Double-Pouch Twist-Seal Bags
PE bags, transparent. These sterile sampling bags provide a secure, contaminant-free pliant container that ensures dependable analysis results. Economical and efficient way to collect, contain and carry samples. With these bags, the user can bring sterile sampling bags into a cleanroom without any worries about contamination. The bags are double-pouched so the user can remove the outer bag before entering the cleanroom, which minimises the number of bags exposed.
- The bags are sterilised inside and out by gamma irradiation
- Sterility certificate in every box
- Lot number traceability for every product
- Maximum opening facilitates inserting samples into the bag
- Easy pull tabs to open the bag without risk of contaminating the interior of the bag
- Bags accommodate solids, semi-solids and liquids
- Free from RNase, DNase, pyrogen and BPA
Regulations: NF ISO 14644-1, ISO 14698, ISO-14644.
: NF ISO 14644-1, ISO 14698, ISO 14644
PE bags, transparent. These sterile sampling bags provide a secure, contaminant-free pliant container that ensures dependable analysis results. Economical and efficient way to collect, contain and carry samples. With these bags, the user can bring sterile sampling bags into a cleanroom without any worries about contamination. The bags are double-pouched so the user can remove the outer bag before entering the cleanroom, which minimises the number of bags exposed.
- The bags are sterilised inside and out by gamma irradiation
- Sterility certificate in every box
- Lot number traceability for every product
- Maximum opening facilitates inserting samples into the bag
- Easy pull tabs to open the bag without risk of contaminating the interior of the bag
- Bags accommodate solids, semi-solids and liquids
- Free from RNase, DNase, pyrogen and BPA
Regulations: NF ISO 14644-1, ISO 14698, ISO-14644.
: NF ISO 14644-1, ISO 14698, ISO 14644
Product Family Options
Safety tabs - clear - closure with 2 round wires
- DescriptionCapacityPackedL×WThickness
- Double wrapping bags with flush closure, sterile, unmoulded body, 2 round thread closure800 ml40 pouches of 25 bags114×305 mm63 µm1 * 1.000 件¥12,218.00
- Double wrapping bags with flush closure, sterile, unmoulded body, 2 round thread closure650 ml40 pouches of 25 bags140×229 mm76 µm1 * 1.000 件¥13,667.00
- Double wrapping bags with flush closure, sterile, unmoulded body, 2 round thread closure1500 ml40 pouches of 25 bags140×382 mm76 µm1 * 1.000 件¥14,695.00
- Double wrapping bags with flush closure, sterile, unmoulded body, 2 round thread closure1650 ml40 pouches of 25 bags178×305 mm76 µm1 * 1.000 件¥15,584.00
Safety tabs - printed - closure with 2 round wires
- DescriptionCapacityPackedL×WThickness
- Double wrapping bags with flush closure, sterile, unmoulded body, 2 round thread closure800 ml40 pouches of 25 bags114×305 mm63 µm1 * 1.000 件¥14,128.00
- Double wrapping bags with flush closure, sterile, printed body, 2 round thread closure650 ml40 pouches of 25 bags140×229 mm76 µm1 * 1.000 件¥14,151.00
- Double wrapping bags with flush closure, sterile, printed body, 2 round thread closure1500 ml40 pouches of 25 bags140×382 mm76 µm1 * 1.000 件¥15,532.00
- Double wrapping bags with flush closure, sterile, printed body, 2 round thread closure1650 ml40 pouches of 25 bags178×305 mm76 µm1 * 1.000 件¥18,084.00
Safety tabs - clear - closure with 1 round wire and 1 flat wire
- DescriptionCapacityPackedL×WThickness
- Double wrapping bags with flush closure, sterile, unmoulded body, closure with 1 round thread and 1 plate650 ml40 pouches of 25 bags140×229 mm63 µm1 * 1.000 件¥13,504.00
- Double wrapping bags with flush closure, sterile, unmoulded body, closure with 1 round thread and 1 plate650 ml40 pouches of 25 bags140×229 mm101 µm1 * 1.000 件¥15,702.00
- Double wrapping bags with flush closure, sterile, unmoulded body, closure with 1 round thread and 1 plate1080 ml40 pouches of 25 bags140×305 mm76 µm1 * 1.000 件¥14,147.00
- Double wrapping bags with flush closure, sterile, unmoulded body, closure with 1 round thread and 1 plate1650 ml40 pouches of 25 bags178×305 mm76 µm1 * 1.000 件¥17,634.00
- Double wrapping bags with flush closure, sterile, unmoulded body, closure with 1 round thread and 1 plate1650 ml40 pouches of 25 bags178×305 mm101 µm1 * 1.000 件¥19,938.00
- Double wrapping bags with flush closure, sterile, printed body, closure with 1 round thread and 1 plate2250 ml40 pouches of 25 bags254×305 mm101 µm1 * 1.000 件¥22,332.00
- Double wrapping bags with flush closure, sterile, unmoulded body, closure with 1 round thread and 1 plate4000 ml40 pouches of 25 bags254×382 mm101 µm1 * 1.000 件¥14,086.00
Safety tabs - printed - closure with 1 round wire and 1 flat wire
- DescriptionCapacityPackedL×WThickness
- Double wrapping bags with flush closure, sterile, printed body, closure with 1 round thread and 1 plate650 ml40 pouches of 25 bags140×229 mm76 µm1 * 1.000 件¥14,710.00
- Double wrapping bags with flush closure, sterile, printed body, closure with 1 round thread and 1 plate650 ml40 pouches of 25 bags140×229 mm101 µm1 * 1.000 件¥15,790.00
- Double wrapping bags with flush closure, sterile, printed body, closure with 1 round thread and 1 plate1080 ml40 pouches of 25 bags140×305 mm76 µm1 * 1.000 件¥16,147.00
- Double wrapping bags with flush closure, sterile, printed body, closure with 1 round thread and 1 plate1650 ml40 pouches of 25 bags178×305 mm76 µm1 * 1.000 件¥19,424.00
- Double wrapping bags with flush closure, sterile, printed body, closure with 1 round thread and 1 plate1650 ml40 pouches of 25 bags178×305 mm101 µm1 * 1.000 件¥20,890.00
- Double wrapping bags with flush closure, sterile, printed body, closure with 1 round thread and 1 plate2250 ml40 pouches of 25 bags254×305 mm101 µm1 * 1.000 件¥21,966.00
- Double wrapping bags with flush closure, sterile, printed body, closure with 1 round thread and 1 plate4000 ml40 pouches of 25 bags254×382 mm101 µm1 * 1.000 件¥25,478.00