Presence-Absence media in bottles
Catalog # HACH2401612
Supplier: HACH
CAS Number:
Specifications
- Pk:12
- Description:Presence-Absence broth with MUG, sterile bottle
- Seq:04
Specifications
About this item
Selective for Total Coliform Bacteria and Escherichia coli. Broth in pre-filled bottles.
- Sterile
- Disposable