About this item
PS, white. These disposable spatulas are ideal for the sampling of powder, granules and pastes.
- Long, ergonomic and solid handle
- Pointed blade can be easily pierced into paper and plastic bags
- Available non sterile (LaboPlast) or gamma radiation sterilised (SteriPlast)
- Production, assembling and packaging acc. to ISO cleanroom class 7
- According to EU food and FDA guidelines
As an option, the spatulas are available with cover in order to secure a contamination-free sampling process.