Electronic pipettes, VIAFLO
Supplier: INTEGRA
Certificates
About this item
The ultra lightweight, well balanced design of the VIAFLO pipettes feature unsurpassed multi-channel pipetting ergonomics, even during prolonged pipetting sessions.
- Effortless tip attachment and ejection forces
- GripTips snap into place with minimal tip loading effort, providing a secure connection
- Colour screen and touch wheel interface offer quick and comprehensive menu navigation
- Ideal for both left and right handed users
All models offer a choice of 10 predefined pipetting modes covering typical applications as well as up to 20 custom multi-step programs for specific tasks. Predefined pipetting modes include: Repetitive dispensing, manual pipetting, sample dilution, reversive pipetting and variable pipetting.
Ordering information: Units are supplied with rechargeable battery (located inside the pipette, Li-ion, 3,7 V), a bag of spare O-rings (300 and 1250 µl volume ranges only), O-ring removal tool (300 and 1250 µl volume ranges only) and accuracy and precision certificate. Supplied without power supply and tips. Those need to be ordered separately.