Reagent reservoirs for multi-channel pipettes and automatic systems
Catalog # BRND701452
Supplier: BRAND
Specifications
- Type:12 columns, low profile, pyramid bottom
- Capacity:12×6 ml
- Colour:White
- Seq:05
- Pk:10
Specifications
About this item
White PP, autoclavable. These reagent reservoirs were designed especially for use with automated liquid handling platforms. They can also be used with manual systems. Different partitions, bottom styles and profiles provide broad application flexibility.
- ANSI/SLAS standard format: Compatible with nearly all liquid handling robots and other automated dispensing systems
- Excellent stability and chemical resistance
- Very low dead volume due to pyramid bottoms
- Free from RNases and DNases
- Autoclavable (121 °C, 20 min)