Order Entry
China
ContactUsLinkComponent
Ion exchange resins, beads, BAKER ANALYZED® IONAC® NM-60 H+/OHˉ Form, Type I 16-50 Mesh (mixed bed exchanger), J.T.Baker®
  BAKR4631-01
 :  Avantor
 :  
undefined
Ion exchange resins, beads, BAKER ANALYZED® IONAC® NM-60 H+/OHˉ Form, Type I 16-50 Mesh (mixed bed exchanger), J.T.Baker®
  BAKR4631-01
 :  Avantor
 :  
  • Pk:
    500 g
  • Seq:
    3645
  • For Laboratory,Research,or Manufacturing Use:
    = 5 %
  • Bead Size Distribution: Greater than 1.250 mm:
    ≤5 %
  • Bead Size Distribution: Less than 0.315 mm:
    ≤2 %
  • Operating capacity /mixed bed at 0.02 MOhm-cm, Eq/L:
    ≥0.55
  • Resistivity/Operating Capacity Mixed Bed test (NaCl): Resistivity after exhaustion, MOhm-cm (in minutes):
    ≥16
  • ¹ Measured to a 20,000 ohm-cm resistivity end point. The unit kgr/cf = kilograins per cubic foot.:
    ≥16
 :