


Ammonia solution 28 - 30% VLSI for the electronics industry, J.T.Baker®
BAKR5358-03
: Avantor
:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Pack type:Poly bottle pressure bottom, bagged
- Pk:8 pt US
- Seq:8026
- For Microelectronic Use:Passes Test
- Appearance:Passes Test
- Assay (as NH₃):28.0 - 30.0 %
- Color (APHA):≤ 10
- Insoluble Matter:≤ 1 ppm
- Residue after Ignition:≤ 3 ppm
- Carbon Dioxide (CO₂):≤ 10 ppm
- Pyridine:Passes Test
- Substances Reducing Permanganate:Passes Test
- Chloride (Cl):≤ 0.3 ppm
- Phosphate (PO₄):≤ 0.20 ppm
- Total Sulfur (as SO₄):≤ 0.8 ppm
- Trace Impurities - Aluminum (Al):≤ 10.0 ppb
- Arsenic and Antimony (as As):≤ 30.0 ppb
- Trace Impurities - Barium (Ba):≤ 10.0 ppb
- Trace Impurities - Beryllium (Be):≤ 10.0 ppb
- Trace Impurities - Bismuth (Bi):≤ 20.0 ppb
- Trace Impurities - Boron (B):≤ 10.0 ppb
- Trace Impurities - Cadmium (Cd):≤ 5.0 ppb
- Trace Impurities - Calcium (Ca):≤ 100.0 ppb
- Trace Impurities - Chromium (Cr):≤ 5.0 ppb
- Trace Impurities - Cobalt (Co):≤ 1.0 ppb
- Trace Impurities - Copper (Cu):≤ 10.0 ppb
- Trace Impurities - Gallium (Ga):≤ 50 ppb
- Trace Impurities - Germanium (Ge):≤ 50.0 ppb
- Trace Impurities - Gold (Au):≤ 10.0 ppb
- Heavy Metals (as Pb):≤ 200.0 ppb
- Trace Impurities - Iron (Fe):≤ 10.0 ppb
- Trace Impurities - Lead (Pb):≤ 10.0 ppb
- Trace Impurities - Lithium (Li):≤ 20.0 ppb
- Trace Impurities - Magnesium (Mg):≤ 50.0 ppb
- Trace Impurities - Manganese (Mn):≤ 5.0 ppb
- Trace Impurities - Mercury (Hg):≤ 5.0 ppb
- Trace Impurities - Molybdenum (Mo):≤ 10.0 ppb
- Trace Impurities - Nickel (Ni):≤ 5.0 ppb
- Trace Impurities - Niobium (Nb):≤ 10.0 ppb
- Trace Impurities - Potassium (K):≤ 100 ppb
- Trace Impurities - Silicon (Si):≤ 150.0 ppb
- Trace Impurities - Silver (Ag):≤ 5.0 ppb
- Trace Impurities - Sodium (Na):≤ 100.0 ppb
- Trace Impurities - Strontium (Sr):≤ 10.0 ppb
- Trace Impurities - Tantalum (Ta):≤ 10.0 ppb
- Trace Impurities - Thallium (Tl):≤ 50.0 ppb
- Trace Impurities - Tin (Sn):≤ 20.0 ppb
- Trace Impurities - Titanium (Ti):≤ 10.0 ppb
- Trace Impurities - Vanadium (V):≤ 20.0 ppb
- Trace Impurities - Zinc (Zn):≤ 5.0 ppb
- Trace Impurities - Zirconium (Zr):≤ 10.0 ppb
- Particle Count - 0.5 µm and greater:≤ 100 par/ml
- Particle Count - 1.0 µm and greater:≤ 10 par/ml
:
:35.05 g/mol
:32…37.7 °C (1013 hPa)
:–60…–57.5 °C
:0.88…0.91 g/cm³ (20 °C)
:Ambient
:215-647-6
:2672
:8,III
:GHS05
:GHS07
:GHS09
:1336-21-6
:7664-41-7
:NH₃
:MFCD00066650