



Mercury(II) sulphate ≥98.0%, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:7783-35-9
:
Formula:HgSO₄
MDL Number:MFCD00011047
:296.65 g/mol
Merck Index:13,05914
Melting Pt:850 °C
Density:6.47 g/cm³ (25 °C)
EINECS:231-992-5
UN:1645
ADR:6.1,II
:GHS06
:GHS08
:GHS09
:GHS07
Storage Temperature:Ambient
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- Meets ACS Reagent Chemical Requirements:≥98.0%
- For Laboratory,Research,or Manufacturing Use:≤0.020%
- Assay (HgSO₄):≥98.0 %
- Residue after Reduction:≤0.020 %
- Chloride (Cl):≤0.003 %
- Nitrate (NO₃):Passes Test
- Trace Impurities - Iron (Fe):≤0.001 %
- Mercurous Mercury (as Hg):≤0.15 %
- Suitability for Chemical Oxygen Demand (COD) Determination:Passes Test