Serological pipettes, PYREX®
Supplier: CORNING SCIENCE PRODUCTS
Certificates
About this item
Reusable serological pipettes made of PYREX® glass.
- Calibrated to deliver (TD)
- Coloured graduations are enameled on to the glass
- Colour-coded by size for easy identification and sorting
Tip and top are tempered to provide increased strength. Pippets are calibrated 'to deliver' and must be blown out to obtain total rated capacity.
Certifications: ASTM E-1044