Volumetric pipettes, class A, color-coded, graduated, serialised/certified, PYREX®
Pipettes are calibrated to Class A tolerances in accordance with ASTM E-542 and ASTM E-969.
- Sizes 1 ml through 25 ml are color-coded for ease in sorting and selection
- Coloured graduations are enamelled on to the glass
PYREX® Serialized/Certified Volumetric Glassware is now tested and calibrated in an ISO/IEC 17025 certified laboratory.
: Each pipette is individually serialised and supplied with a certificate of identity and capacity, traceable to NIST standards.