Order Entry
China
ContactUsLinkComponent
Barium sulphate ≥97.0%, powder, BAKER ANALYZED®, J.T.Baker®
Barium sulphate ≥97.0%, powder, BAKER ANALYZED®, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Barium sulphate ≥97.0%, powder, BAKER ANALYZED®, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    ≥97.0%
  • Assay (BaSO₄):
    ≥97.0 %
  • Acidity (as H₂SO₄):
    ≤0.01 %
  • Water-Soluble Salts:
    ≤0.02 %
  • Chloride (Cl):
    ≤0.005 %
  • Phosphate (PO₄):
    ≤0.001 %
  • Heavy Metals (as Pb):
    ≤0.001 %
  • Trace Impurities - Iron (Fe):
    ≤0.001 %
  • Trace Impurities - Arsenic (As):
    ≤1 ppm

Specification Test Results

Related Information