


Acetic acid glacial ≥99.7% (by GC, corrected for water content), BAKER ANALYZED® ACS aldehyde-free, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:64-19-7
:
Formula:H₃CCOOH
MDL Number:MFCD00036152
:60.05 g/mol
Boiling Pt:118 °C (1013 hPa)
Melting Pt:17 °C
Density:1.05 g/cm³ (20 °C)
Flash Pt:38.5 °C
Storage Temperature:Ambient
EINECS:200-580-7
UN:2789
ADR:8,II
REACH:01-2119475328-30
:GHS02
:GHS05
:GHS07
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:= 99.7 %
- IMPORTANT: Material will freeze if stored below 17 °C (63°F).:= 10
- Assay (by GC, corrected for water):≥99.7 %
- Color (APHA):≤10
- Dilution Test:Passes Test
- Residue after Evaporation:≤0.001 %
- Acetic Anhydride ((CH₃CO)₂O):≤0.01 %
- Chloride (Cl):≤1 ppm
- Sulfate (SO₄):≤1 ppm
- Trace Impurities - Iron (Fe):≤0.2 ppm
- Substances Reducing Dichromate:Passes Test
- Substances Reducing Permanganate:Passes Test
- Titrable Base (meq/g):≤0.0004
- Acetaldehyde:≤0.005 %
- Sensitivity:Passes Test
- Trace Impurities - Arsenic and Antimony (as As):≤50.0 ppb
- Trace Impurities - Boron (B):≤100.0 ppb
- Trace Impurities - Chromium (Cr):≤200.0 ppb
- Trace Impurities - Cobalt (Co):≤100.0 ppb
- Trace Impurities - Copper (Cu):≤100.0 ppb
- Trace Impurities - Heavy Metals:≤0.5 ppm
- Trace Impurities - Lead (Pb):≤300.0 ppb
- Trace Impurities - Manganese (Mn):≤200.0 ppb
- Trace Impurities - Nickel (Ni):≤100.0 ppb
- Trace Impurities - Potassium (K):≤300.0 ppb
- Trace Impurities - Tin (Sn):≤100 ppb
- Trace Impurities - Titanium (Ti):≤300.0 ppb
- Trace Impurities - Zinc (Zn):≤200.0 ppb
Specification Test Results
Related Information
Certificates
Search by Lot Numberspec_BAKR9508-03 spec_BAKR9508-07 spec_BAKR9508-15