硫酸铝钾 98.0-102.0% (通过EDTA滴定), crystals, BAKER ANALYZED® ACS, J.T.Baker®
: Avantor
:7784-24-9
:
:AlK(SO₄)₂·12H₂O
:MFCD00149143
:474.39 g/mol
:12,00373
:1,72 g/cm³ (20 °C)
:233-141-3
- Meets ACS Reagent Chemical Requirements:98.0 - 102.0%
- For Laboratory,Research,or Manufacturing Use:99.0 - 100.5%
- Assay (AlK(SO₄)₂·12H₂O) (by EDTA titrn):98.0 - 102.0 %
- Assay (dried basis):99.0 - 100.5 %
- Insoluble Matter:≤0.005 %
- pH of 5% Solution at 25°C:2.0 - 4.0
- Ammonium (NH₄):≤0.005 %
- Trace Impurities - Iron (Fe):≤0.001 %
- Sodium (Na):≤0.02 %
- Chloride (Cl):≤5 ppm
- Trace Impurities - ACS - Heavy Metals (as Pb):≤5 ppm