Order Entry
China
ContactUsLinkComponent
VWR®, Disposal Box for Pipette Tips
VWR®, Disposal Box for Pipette Tips
Catalog # VWRI212-1833
Supplier:  VWR Collection
undefined
VWR®, Disposal Box for Pipette Tips
Catalog # VWRI212-1833
Supplier:  VWR Collection
Specifications
  • Description:
    Disposal box for pipette tips, clear acrylic, with flip cover
  • W×D×H:
    154×171×206 mm
  • Pk:
    1
Specifications
About this item
This acrylic box is ideal for the one-handed disposal of used pipette tips. The 10 mm thick acrylic provides protection from β radiation.
  • Quick flip of port cover makes tip ejection easy
  • Eliminates any need to handle pipette tips used with hazardous fluids
  • Large hinged lid allows easy disposal of contents
W×D×H: 154×171×206 mm