Gel documentation systems, Gel Doc™ XR+Molecular Imager®
BRAD170-8195
: BIO RAD
- Resolution:4 megapixels
- Description:Gel Documentation Systems, Gel Doc™ XR+ Molecular Imager®
- W×D×H:600×360×900 mm
- Pk:1
Gel Doc™ XR+systems are suitable for laboratories engaged in RNA analysis, molecular diagnostics, epigenetics, pharmacogenomics, forensic testing of genomic DNA.
- Automated workflow
- Auto camera aperture control
- High image resolution, high export functionality
- Auto focus